Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-11
Entrez Gene ID
171952
Gene Name
protein-coding
Gene Aliases
CELE_C32E8.10
Chromosome Location
Chr.I: 3798260 - 3805739 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025843.3 NP_001021014.1 3 - 4 516 95
NM_001025845.4 NP_001021016.1 3 - 4 516 95
NM_001375250.2 NP_001361962.1 3 - 4 518 95
NM_001375251.3 NP_001361960.1 3 - 4 516 95
NM_001375252.3 NP_001361961.1 3 - 4 518 95
NM_058826.2 NP_491227.1 3 - 4 516 95
NM_058829.5 NP_491230.1 3 - 4 501 95
GenBank mRNA AF144257.1 - 3 - 4 483 95
AF144258.1 - 3 - 4 483 95
AF144259.1 - 3 - 4 483 95

View Full Product Details