Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cas-2
Entrez Gene ID
266829
Gene Name
protein-coding
Gene Aliases
CELE_C18E3.6
Chromosome Location
Chr.I: 4185176 - 4193132 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306365.3 NP_001293294.1 1 - 2 544 150
NM_058923.7 NP_491324.1 5 - 6 1090 150
NR_131381.1 5 - 6 986 150

View Full Product Details