Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
anc-1
Entrez Gene ID
172034
Gene Name
protein-coding
Gene Aliases
CELE_ZK973.6
Chromosome Location
Chr.I: 4318033 - 4359092 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356817.3 NP_001343707.1 16 - 17 4352 70
NM_001356818.3 NP_001343735.1 5 - 6 1444 70
NM_058952.5 NP_491353.2 16 - 17 4373 70

View Full Product Details