Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
app-1
Entrez Gene ID
172118
Gene Name
Xaa-Pro aminopeptidase app-1
Gene Aliases
CELE_W03G9.4
Chromosome Location
Chr.I: 4964623 - 4967211 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059088.7 NP_491489.1 3 - 4 592 129

View Full Product Details