Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phat-1
Entrez Gene ID
172129
Gene Name
PHAryngeal gland Toxin-related
Gene Aliases
CELE_C46H11.8
Chromosome Location
Chr.I: 5044198 - 5045720 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059107.3 NP_491508.2 2 - 3 155 64

View Full Product Details