Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-16
Entrez Gene ID
172134
Gene Name
V-type proton ATPase subunit
Gene Aliases
CELE_C30F8.2
Chromosome Location
Chr.I: 5079910 - 5082273 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059114.7 NP_491515.2 3 - 4 487 77

View Full Product Details