Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apm-1
Entrez Gene ID
172180
Gene Name
MHD domain-containing protein
Gene Aliases
CELE_F55A12.7
Chromosome Location
Chr.I: 5342161 - 5344724 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059171.8 NP_491572.2 1 - 2 65 76

View Full Product Details