Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
obr-4
Entrez Gene ID
172253
Gene Name
protein-coding
Gene Aliases
CELE_C32F10.1
Chromosome Location
Chr.I: 5829391 - 5834642 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380388.1 NP_001367357.1 3 - 4 999 138
NM_059290.7 NP_491691.1 8 - 9 1836 138

View Full Product Details