Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
avr-14
Entrez Gene ID
172270
Gene Name
protein-coding
Gene Aliases
CELE_B0207.12
Chromosome Location
Chr.I: 5967245 - 5973999 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383179.1 6 - 7 716 106

View Full Product Details