Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pmk-2
Entrez Gene ID
177611
Gene Name
protein-coding
Gene Aliases
CELE_F42G8.3
Chromosome Location
Chr.IV: 8143500 - 8148648 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380352.1 NP_001368440.1 2 - 3 148 77
NM_001380353.1 NP_001368441.1 2 - 3 148 77
NM_171392.7 NP_741457.2 7 - 8 768 77
NM_171913.6 NP_741458.2 7 - 8 757 77

View Full Product Details