Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- F20C5.11
- Entrez Gene ID
-
36805005
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F20C5.11
- Chromosome Location
-
Chr.IV: 8755035 - 8762076 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383152.1 |
|
9 - 10 |
1890 |
134 |
- Gene Symbol
- klp-11
- Entrez Gene ID
-
177685
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F20C5.2
- Chromosome Location
-
Chr.IV: 8755045 - 8762066 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383153.1 |
|
9 - 10 |
1885 |
134 |
View Full Product Details