Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-34
Entrez Gene ID
177800
Gene Name
protein-coding
Gene Aliases
CELE_F58G6.5
Chromosome Location
Chr.IV: 9664361 - 9675672 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028074.5 NP_001023245.1 7 - 8 1284 97
NM_001136387.4 NP_001129859.1 5 - 6 1071 97
NM_069315.6 NP_501716.2 7 - 8 1237 97
NM_171412.4 NP_741480.1 5 - 6 940 97

View Full Product Details