Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rde-1
Entrez Gene ID
179393
Gene Name
Piwi domain-containing protein
Gene Aliases
CELE_K08H10.7
Chromosome Location
Chr.V: 9987916 - 9991644 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171525.6 NP_741611.1 4 - 5 655 90
GenBank mRNA AF180730.1 - 4 - 5 645 90

View Full Product Details