Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sma-1
Entrez Gene ID
179629
Gene Name
protein-coding
Gene Aliases
CELE_R31.1
Chromosome Location
Chr.V: 11899449 - 11916623 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136431.4 NP_001129903.1 6 - 7 4519 74
NM_001269453.3 NP_001256382.1 4 - 5 4270 74
NM_001269454.2 NP_001256383.1 6 - 7 4761 74
NM_001373124.2 NP_001360523.1 7 - 7 4828 74
NM_001383415.1 6 - 7 4767 74
NM_171544.7 NP_741632.2 7 - 8 4943 74
GenBank mRNA AF053496.1 - 6 - 7 4519 74

View Full Product Details