Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgef-1
Entrez Gene ID
180571
Gene Name
protein-coding
Gene Aliases
CELE_C14A11.3
Chromosome Location
Chr.X: 2772042 - 2799250 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171642.6 NP_741737.1 5 - 6 2454 94

View Full Product Details