Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-65
Entrez Gene ID
180734
Gene Name
protein-coding
Gene Aliases
CELE_T13H2.4
Chromosome Location
Chr.X: 4610911 - 4619635 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375019.2 NP_001361932.1 6 - 6 776 78
NM_001375020.2 NP_001362134.1 5 - 6 774 78
NM_001375021.2 NP_001361933.1 8 - 9 2324 78
NM_001375022.2 NP_001362133.1 8 - 9 2326 78
NM_001375024.2 NP_001362136.1 7 - 8 2251 78
NM_001383564.1 8 - 9 2322 78

View Full Product Details