Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-65
Entrez Gene ID
180734
Gene Name
protein-coding
Gene Aliases
CELE_T13H2.4
Chromosome Location
Chr.X: 4610911 - 4619635 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375021.2 NP_001361933.1 3 - 4 253 74
NM_001375022.2 NP_001362133.1 3 - 4 249 74
NM_001375024.2 NP_001362136.1 2 - 3 174 74

View Full Product Details