Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chtl-1
Entrez Gene ID
180809
Gene Name
protein-coding
Gene Aliases
CELE_F35C8.7
Chromosome Location
Chr.X: 5355637 - 5364322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380966.2 NP_001367416.1 3 - 4 724 96
NM_001380967.2 NP_001367417.1 3 - 4 102
NM_171686.7 NP_741790.2 5 - 6 626 96

View Full Product Details