Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kqt-1
Entrez Gene ID
180933
Gene Name
protein-coding
Gene Aliases
CELE_C25B8.1
Chromosome Location
Chr.X: 6619012 - 6627995 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171709.6 NP_741819.1 11 - 12 1289 88
NM_171710.7 NP_741820.1 10 - 11 1304 88
GenBank mRNA AY572974.1 - 10 - 11 1135 88

View Full Product Details