Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kqt-1
Entrez Gene ID
180933
Gene Name
protein-coding
Gene Aliases
CELE_C25B8.1
Chromosome Location
Chr.X: 6619012 - 6627995 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171709.6 NP_741819.1 1 - 2 113 73

View Full Product Details