Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sma-5
Entrez Gene ID
181373
Gene Name
protein-coding
Gene Aliases
CELE_W06B3.2
Chromosome Location
Chr.X: 11997515 - 12005549 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047832.3 NP_001041297.1 7 - 8 1278 109
NM_001381114.2 NP_001366649.1 7 - 8 1823 109
NM_001383653.1 7 - 8 1305 109
NM_171782.6 NP_741908.1 6 - 7 1253 109

View Full Product Details