Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-50
Entrez Gene ID
181413
Gene Name
protein-coding
Gene Aliases
CELE_T07C5.1
Chromosome Location
Chr.X: 12691566 - 12695652 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313450.3 NP_001300379.1 1 - 2 67 111
NM_001313451.3 NP_001300380.1 2 - 2 475 111
NM_001373541.1 NP_001359531.1 2 - 3 528 111
NM_171786.7 NP_741913.1 2 - 3 535 111

View Full Product Details