Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-32
Entrez Gene ID
176257
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.8
Chromosome Location
Chr.III: 8905615 - 8911756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027846.2 NP_001023017.1 1 - 2 169 89
NM_001027847.4 NP_001023018.1 1 - 2 169 89
NM_001027848.6 NP_001023019.1 1 - 2 169 89
NM_001027849.4 NP_001023020.1 1 - 2 169 89
NM_001027850.5 NP_001023021.1 1 - 2 169 89
NM_001027851.2 NP_001023022.1 1 - 2 169 89
GenBank mRNA AF320899.1 - 1 - 2 167 89
AF320900.1 - 1 - 2 167 89
AF320901.1 - 1 - 2 167 89
AF320902.1 - 1 - 2 167 89
AF320903.1 - 1 - 2 167 89
AF320904.1 - 1 - 2 167 89

View Full Product Details