Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pde-4
Entrez Gene ID
174235
Gene Name
protein-coding
Gene Aliases
CELE_R153.1
Chromosome Location
Chr.II: 7627245 - 7653856 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047333.6 NP_001040798.1 10 - 11 1538 74
NM_001306543.3 NP_001293472.1 10 - 11 1391 74
NM_001383892.1 10 - 11 1451 74
NM_063199.6 NP_495600.1 10 - 11 1473 74
NM_063200.4 NP_495601.1 8 - 9 1571 74
NM_182144.6 NP_871944.1 11 - 12 2151 74
NM_182145.5 NP_871945.1 8 - 9 1644 74

View Full Product Details