Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abts-3
Entrez Gene ID
174024
Gene Name
protein-coding
Gene Aliases
CELE_F57F10.1
Chromosome Location
Chr.II: 6132895 - 6157205 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368589.3 NP_001355398.1 10 - 11 1193 93
NM_001368590.4 NP_001355414.1 - 4495 93
NM_001381447.1 NP_001367591.1 9 - 10 1231 93
NM_001381448.2 NP_001367590.1 9 - 10 1242 93
NM_062827.5 NP_495228.1 8 - 9 1489 93
GenBank mRNA AY887905.1 - 8 - 9 1511 93

View Full Product Details