Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-69
Entrez Gene ID
353477
Gene Name
Serpentine receptor class gamma-69
Gene Aliases
CELE_F09E5.4
Chromosome Location
Chr.II: 5355684 - 5357361 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377800.1 NP_001364743.1 4 - 5 573 76

View Full Product Details