Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-10
Entrez Gene ID
173021
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_Y53H1B.1
Chromosome Location
Chr.I: 11294576 - 11297206 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060499.7 NP_492900.1 4 - 5 470 69

View Full Product Details