Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-34
Entrez Gene ID
173024
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F56G4.1
Chromosome Location
Chr.I: 11342085 - 11347288 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060507.4 NP_492908.2 9 - 10 1745 77

View Full Product Details