Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-11
Entrez Gene ID
188765
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_T22H2.3
Chromosome Location
Chr.I: 11694577 - 11697943 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060578.3 NP_492979.2 5 - 6 820 105

View Full Product Details