Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-59
Entrez Gene ID
173233
Gene Name
Septin-type G domain-containing protein
Gene Aliases
CELE_W09C5.2
Chromosome Location
Chr.I: 13629227 - 13633509 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060987.5 NP_493388.1 2 - 3 381 78

View Full Product Details