Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ins-30
Entrez Gene ID
173267
Gene Name
INSulin related
Gene Aliases
CELE_ZC334.2
Chromosome Location
Chr.I: 14034954 - 14036057 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061043.4 NP_493444.1 1 - 2 283 58

View Full Product Details