Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fipr-24
Entrez Gene ID
183286
Gene Name
FIP (Fungus-Induced Protein) Related
Gene Aliases
CELE_C37A5.8
Chromosome Location
Chr.I: 14157611 - 14158078 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061070.2 NP_493471.1 1 - 2 199 80

View Full Product Details