Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lev-11
Entrez Gene ID
173319
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.1
Chromosome Location
Chr.I: 14620814 - 14631254 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026526.6 NP_001021697.1 6 - 7 689 78
NM_001381261.2 NP_001368155.1 8 - 9 800 78
NM_001381263.2 NP_001367007.1 5 - 6 509 78
NM_001381264.1 NP_001368156.1 5 - 6 499 78
NM_001381265.1 NP_001368154.1 4 - 5 385 78
NM_001383761.1 8 - 9 907 78
NM_001383762.1 6 - 7 689 78
GenBank mRNA AF298180.1 - 6 - 7 688 78
D38541.1 - 8 - 9 836 78
LC215398.1 - 6 - 7 688 78

View Full Product Details