Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lev-11
Entrez Gene ID
173319
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.1
Chromosome Location
Chr.I: 14620814 - 14631254 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026526.6 NP_001021697.1 2 - 3 290 111
NM_001313525.4 NP_001300454.1 2 - 3 301 111
GenBank mRNA AF298180.1 - 2 - 3 289 111

View Full Product Details