Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lev-11
Entrez Gene ID
173319
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8B.1
Chromosome Location
Chr.I: 14620814 - 14631254 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383763.1 6 - 7 693 61
GenBank mRNA D38542.1 - 6 - 7 693 61

View Full Product Details