Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
uev-1
Entrez Gene ID
173347
Gene Name
Ubiquitin-conjugating enzyme E2 variant 1
Gene Aliases
CELE_F39B2.2
Chromosome Location
Chr.I: 14790244 - 14790934 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061177.6 NP_493578.1 2 - 3 175 72

View Full Product Details