Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-3
Entrez Gene ID
187251
Gene Name
Serpentine receptor class delta-3
Gene Aliases
CELE_K10B4.5
Chromosome Location
Chr.II: 126779 - 129029 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061266.2 NP_493667.1 2 - 3 368 150

View Full Product Details