Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snt-6
Entrez Gene ID
182427
Gene Name
protein-coding
Gene Aliases
CELE_C08G5.4
Chromosome Location
Chr.II: 745418 - 750212 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267093.2 NP_001254022.1 6 - 7 847 79
NM_001267094.2 NP_001254023.1 3 - 4 487 79
NM_001318279.3 NP_001305208.1 2 - 2 271 79

View Full Product Details