Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-34
Entrez Gene ID
187667
Gene Name
F-box C protein
Gene Aliases
CELE_R07C3.5
Chromosome Location
Chr.II: 931171 - 932327 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061454.5 NP_493855.2 2 - 3 787 93

View Full Product Details