Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lips-11
Entrez Gene ID
188467
Gene Name
LIPaSe related
Gene Aliases
CELE_T13B5.5
Chromosome Location
Chr.II: 1108841 - 1111001 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061513.2 NP_493914.1 3 - 4 705 75

View Full Product Details