Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-31
Entrez Gene ID
190362
Gene Name
GLoBin related
Gene Aliases
CELE_Y57G7A.9
Chromosome Location
Chr.II: 1300064 - 1301654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061543.4 NP_493944.1 2 - 3 409 122

View Full Product Details