Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-54
Entrez Gene ID
173529
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_K05F6.7
Chromosome Location
Chr.II: 1555213 - 1556335 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061594.2 NP_493995.1 1 - 2 116 121

View Full Product Details