Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-38
Entrez Gene ID
188249
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_T07H3.3
Chromosome Location
Chr.II: 1577378 - 1579986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061600.5 NP_494001.3 4 - 5 1207 138

View Full Product Details