Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-22
Entrez Gene ID
173538
Gene Name
F-box C protein
Gene Aliases
CELE_F58E1.10
Chromosome Location
Chr.II: 1688169 - 1691111 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061642.6 NP_494043.2 1 - 2 87 63

View Full Product Details