Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-4
Entrez Gene ID
173548
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_C08F1.5
Chromosome Location
Chr.II: 1778772 - 1782574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047270.4 NP_001040735.2 6 - 7 1574 79

View Full Product Details