Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-7
Entrez Gene ID
182666
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_C16C4.12
Chromosome Location
Chr.II: 1885439 - 1886479 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061709.1 NP_494110.1 1 - 2 98 106

View Full Product Details