Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-42
Entrez Gene ID
173566
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_T16A1.1
Chromosome Location
Chr.II: 2098871 - 2104180 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377713.3 NP_001364517.1 3 - 4 578 142

View Full Product Details