Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-74
Entrez Gene ID
191930
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_F33H12.4
Chromosome Location
Chr.II: 2580820 - 2583346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061898.3 NP_494299.1 5 - 6 1195 82

View Full Product Details