Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-36
Entrez Gene ID
191910
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_F33H12.5
Chromosome Location
Chr.II: 2583732 - 2585593 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061899.2 NP_494300.1 1 - 2 150 93

View Full Product Details