Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-5
Entrez Gene ID
190083
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_Y49F6B.11
Chromosome Location
Chr.II: 3512690 - 3514731 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062164.2 NP_494565.2 4 - 5 854 121

View Full Product Details